Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037727.2 | NP_001032816.1 | 9 - 10 | 1571 | 60 | |
NM_001286610.1 | NP_001273539.1 | 8 - 9 | 1371 | 60 | ||
NM_175476.4 | NP_780685.2 | 9 - 10 | 1242 | 60 | ||
NR_104483.1 | 9 - 10 | 1705 | 60 | |||
GenBank mRNA | AK037710.1 | - | 9 - 10 | 1304 | 60 | |
AK048450.1 | - | 9 - 10 | 1202 | 60 | ||
AK137444.1 | - | 9 - 10 | 1230 | 60 | ||
AK137526.1 | - | 9 - 10 | 1642 | 60 | ||
AK159598.1 | - | 8 - 9 | 1372 | 60 | ||
AK159941.1 | - | 8 - 9 | 1372 | 60 | ||
AK172878.1 | - | 7 - 8 | 901 | 60 | ||
BC108400.1 | - | 6 - 7 | 852 | 60 | ||
BC138751.1 | - | 8 - 9 | 1150 | 8861773 | 60 | |
BC138752.1 | - | 8 - 9 | 1150 | 8861774 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |