Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033253.3 | NP_001028425.1 | 3 - 4 | 674 | 106 | |
NM_001159942.1 | NP_001153414.1 | 4 - 5 | 840 | 106 | ||
GenBank mRNA | AK033676.1 | - | 4 - 5 | 840 | 106 | |
AK034403.1 | - | 4 - 5 | 841 | 106 | ||
AK045140.1 | - | 4 - 5 | 1020 | 106 | ||
AK135923.1 | - | 3 - 4 | 676 | 106 | ||
BC076628.1 | - | 1 - 2 | 71 | 106 | ||
BC117965.1 | - | 2 - 3 | 276 | 40098326 | 106 | |
BC117966.1 | - | 2 - 3 | 269 | 40098330 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |