Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033253.3 | NP_001028425.1 | 7 - 8 | 1074 | 76 | |
NM_001159942.1 | NP_001153414.1 | 8 - 9 | 1240 | 76 | ||
GenBank mRNA | AK034403.1 | - | 8 - 9 | 1241 | 76 | |
AK122468.1 | - | 4 - 5 | 398 | 76 | ||
AK135923.1 | - | 7 - 8 | 1076 | 76 | ||
BC076628.1 | - | 5 - 6 | 471 | 76 | ||
BC117965.1 | - | 6 - 7 | 676 | 40098326 | 76 | |
BC117966.1 | - | 6 - 7 | 669 | 40098330 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |