Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023267.mm.1 |
TC0600003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023267.mm.1 |
NM_009525.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023267.mm.1 |
NM_009525.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023261.mm.1 |
TC0600003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023261.mm.1 |
NM_001271757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023261.mm.1 |
NM_001271758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023261.mm.1 |
NM_009525.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023261.mm.1 |
NM_009525.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023268.mm.1 |
TC0600003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023268.mm.1 |
NM_009525.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023268.mm.1 |
NM_009525.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023262.mm.1 |
TC0600003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023262.mm.1 |
NM_001271757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023262.mm.1 |
NM_001271758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023262.mm.1 |
NM_009525.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023262.mm.1 |
NM_009525.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023269.mm.1 |
TC0600003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023269.mm.1 |
NM_009525.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023269.mm.1 |
NM_009525.3 |
Refseq |