Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009779.2 | NP_033909.1 | 1 - 2 | 178 | 65 | |
GenBank mRNA | AK154134.1 | - | 1 - 2 | 104 | 65 | |
AK154229.1 | - | 1 - 2 | 166 | 65 | ||
AK154609.1 | - | 1 - 2 | 105 | 65 | ||
BC003728.1 | - | 1 - 2 | 91 | 3593758 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |