Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042623.2 | NP_001036088.1 | 10 - 11 | 2331 | 71 | |
NM_001271579.1 | NP_001258508.1 | 11 - 12 | 2458 | 71 | ||
NM_007905.3 | NP_031931.2 | 11 - 12 | 2487 | 71 | ||
NR_073364.1 | 11 - 12 | 2494 | 71 | |||
GenBank mRNA | AK036370.1 | - | 11 - 12 | 2447 | 71 | |
AK132756.1 | - | 10 - 11 | 2289 | 71 | ||
AK167590.1 | - | 10 - 11 | 2320 | 71 | ||
BC046535.1 | - | 11 - 12 | 2474 | 4935713 | 71 | |
BC052394.1 | - | 11 - 12 | 2820 | 5719837 | 71 | |
S73882.1 | - | 11 - 12 | 2449 | 71 | ||
S73883.1 | - | 11 - 12 | 2456 | 71 | ||
S73884.1 | - | 10 - 11 | 2697 | 71 | ||
U63386.1 | - | 10 - 11 | 2415 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |