Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199311.2 | NP_955015.1 | 2 - 3 | 337 | 103 | |
GenBank mRNA | AY397675.1 | - | 2 - 3 | 272 | 103 | |
BC049354.1 | - | 2 - 3 | 327 | 6310008 | 103 | |
CK626014.1 | - | 2 - 3 | 337 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |