Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175628.3 | NP_783327.2 | 18 - 19 | 2357 | 69 | |
GenBank mRNA | AY185125.1 | - | 18 - 19 | 2303 | 69 | |
BC023844.1 | - | 8 - 9 | 1019 | 69 | ||
BC072642.1 | - | 18 - 19 | 2297 | 6839761 | 69 | |
CN701262.1 | - | 1 - 2 | 74 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |