Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199033.1 | NP_950198.1 | 7 - 8 | 993 | 69 | |
GenBank mRNA | AK134697.1 | - | 7 - 8 | 989 | 69 | |
BC061473.1 | - | 7 - 8 | 993 | 6414790 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |