Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145383.1 | NP_663358.1 | 2 - 3 | 607 | 109 | |
GenBank mRNA | AK044333.1 | - | 2 - 3 | 627 | 109 | |
AK044412.1 | - | 2 - 3 | 625 | 109 | ||
BC013125.1 | - | 2 - 3 | 607 | 4500760 | 109 | |
BC031766.1 | - | 2 - 3 | 608 | 4527040 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |