Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029780.3 | NP_084056.1 | 11 - 12 | 1624 | 72 | |
GenBank mRNA | AB057655.1 | - | 10 - 11 | 1169 | 72 | |
AB057663.2 | - | 11 - 12 | 1394 | 72 | ||
AK018272.1 | - | 2 - 3 | 180 | 72 | ||
AK141745.1 | - | 11 - 12 | 1625 | 72 | ||
AK148327.1 | - | 533 | 72 | |||
BC015273.1 | - | 11 - 12 | 1503 | 4222659 | 72 | |
BC056439.1 | - | 11 - 12 | 1541 | 72 | ||
BC092040.1 | - | 11 - 12 | 1489 | 2645313 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |