Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001179.3 | NP_001001179.2 | 28 - 29 | 3596 | 84 | |
GenBank mRNA | AK138836.1 | - | 28 - 29 | 3597 | 84 | |
AK161068.1 | - | 15 - 16 | 1946 | 84 | ||
AK161184.1 | - | 28 - 29 | 3600 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |