Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001179.3 | NP_001001179.2 | 5 - 6 | 575 | 105 | |
GenBank mRNA | AK138836.1 | - | 5 - 6 | 576 | 105 | |
AK161184.1 | - | 5 - 6 | 578 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |