Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK146408.1 | - | 2 - 3 | 887 | 112 | |
AK220217.1 | - | 2 - 3 | 1069 | 112 | ||
BC141065.1 | - | 3 - 4 | 1073 | 9055677 | 112 | |
BC145318.1 | - | 3 - 4 | 1073 | 9053873 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |