Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026849.2 | NP_081125.2 | 1 - 2 | 282 | 65 | |
GenBank mRNA | AK034867.1 | - | 1 - 2 | 229 | 65 | |
AK161445.1 | - | 1 - 2 | 275 | 65 | ||
BC018294.1 | - | 1 - 2 | 232 | 4021243 | 65 | |
CX207820.1 | - | 1 - 2 | 282 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |