Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 19 - 20 | 3311 | 145 | |
GenBank mRNA | AK129082.1 | - | 19 - 20 | 3271 | 145 | |
AK162178.1 | - | 19 - 20 | 3369 | 145 | ||
BC030906.1 | - | 12 - 13 | 2111 | 145 | ||
BC057054.1 | - | 19 - 20 | 3311 | 6827225 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |