Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136227.1 | NP_001129699.1 | 4 - 5 | 565 | 86 | |
NM_009106.2 | NP_033132.2 | 5 - 6 | 972 | 86 | ||
NM_133641.2 | NP_598396.2 | 4 - 5 | 1476 | 86 | ||
GenBank mRNA | AK076155.1 | - | 4 - 5 | 486 | 86 | |
BC004824.1 | - | 4 - 5 | 806 | 86 | ||
BC013820.1 | - | 4 - 5 | 1450 | 3981927 | 86 | |
U54638.1 | - | 5 - 6 | 975 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |