Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007517.4 | NP_031543.2 | 3 - 4 | 596 | 94 | |
NR_125891.1 | 3 - 4 | 596 | 94 | |||
GenBank mRNA | AK154707.1 | - | 3 - 4 | 465 | 94 | |
AK158035.1 | - | 3 - 4 | 387 | 94 | ||
AK167335.1 | - | 3 - 4 | 386 | 94 | ||
AK171011.1 | - | 3 - 4 | 399 | 94 | ||
AK171184.1 | - | 3 - 4 | 391 | 94 | ||
AK171202.1 | - | 3 - 4 | 405 | 94 | ||
AK171249.1 | - | 3 - 4 | 385 | 94 | ||
AK171541.1 | - | 3 - 4 | 386 | 94 | ||
AK171621.1 | - | 3 - 4 | 378 | 94 | ||
AK195286.1 | - | 1 - 2 | 47 | 94 | ||
AK209976.1 | - | 1 - 2 | 47 | 94 | ||
BC016485.1 | - | 3 - 4 | 389 | 3962982 | 94 | |
U41736.1 | - | 3 - 4 | 394 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |