Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023790.2 | NP_076279.1 | 2 - 3 | 431 | 79 | |
GenBank mRNA | AF053628.1 | - | 2 - 3 | 419 | 79 | |
AK006536.1 | - | 2 - 3 | 331 | 79 | ||
AK154345.1 | - | 2 - 3 | 422 | 79 | ||
AK202399.1 | - | 1 - 2 | 46 | 79 | ||
BC083332.1 | - | 3 - 4 | 336 | 6819838 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |