Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033606.3 | NP_291084.1 | 6 - 7 | 1516 | 78 | |
GenBank mRNA | AF318278.1 | - | 5 - 6 | 1160 | 78 | |
BC048936.1 | - | 1 - 2 | 179 | 78 | ||
BC075680.1 | - | 6 - 7 | 1516 | 30612145 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |