Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175277.4 | NP_780486.1 | 1 - 2 | 70 | 60 | |
GenBank mRNA | AK028102.1 | - | 1 - 2 | 71 | 60 | |
AK131950.1 | - | 79 | 60 | |||
AK160488.1 | - | 1 - 2 | 74 | 60 | ||
BC116947.1 | - | 1 - 2 | 108 | 40126266 | 60 | |
BC116949.1 | - | 1 - 2 | 108 | 40126268 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |