Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029245.3 | NP_083521.2 | 4 - 5 | 916 | 84 | |
GenBank mRNA | AK016222.1 | - | 3 - 4 | 712 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |