Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012003.2 | NP_036133.1 | 1 - 2 | 109 | 53 | |
GenBank mRNA | AF071316.1 | - | 1 - 2 | 111 | 53 | |
BG085353.2 | - | 1 - 2 | 109 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |