Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016845.2 | NP_058541.2 | 9 - 10 | 1588 | 68 | |
GenBank mRNA | AK132705.1 | - | 9 - 10 | 1556 | 68 | |
D17573.1 | - | 9 - 10 | 1540 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons |