Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207237.3 | NP_997120.1 | 10 - 11 | 2455 | 74 | |
GenBank mRNA | AK048880.1 | - | 5 - 6 | 697 | 74 | |
AK144486.1 | - | 9 - 10 | 2229 | 74 | ||
AK198104.1 | - | 1 - 2 | 50 | 74 | ||
BC030443.1 | - | 4 - 5 | 618 | 74 | ||
BC067023.1 | - | 11 - 12 | 2025 | 6848230 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |