Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207237.3 | NP_997120.1 | 5 - 6 | 1738 | 93 | |
GenBank mRNA | AK141122.1 | - | 5 - 6 | 1216 | 93 | |
BC067023.1 | - | 6 - 7 | 1308 | 6848230 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |