Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162970.1 | NP_001156442.1 | 8 - 9 | 3184 | 63 | |
GenBank mRNA | AK145690.1 | - | 8 - 9 | 3190 | 63 | |
BC042709.1 | - | 3 - 4 | 205 | 63 | ||
BC150738.1 | - | 7 - 8 | 3113 | 9087649 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |