Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081047.1 | NP_001074516.1 | 10 - 11 | 68 | ||
GenBank mRNA | AK145910.1 | - | 10 - 11 | 68 | ||
BC113805.1 | - | 10 - 11 | 968 | 40062208 | 68 | |
BC113956.1 | - | 10 - 11 | 968 | 40062209 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |