Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011270.3 | NP_035400.3 | 3 - 4 | 547 | 73 | |
GenBank mRNA | AB015189.1 | - | 3 - 4 | 482 | 73 | |
AF012430.1 | - | 3 - 4 | 462 | 73 | ||
AF047827.1 | - | 3 - 4 | 482 | 73 | ||
AF057524.1 | - | 3 - 4 | 499 | 73 | ||
AF069311.1 | - | 3 - 4 | 501 | 73 | ||
AK079335.1 | - | 3 - 4 | 491 | 73 | ||
AK157850.1 | - | 3 - 4 | 474 | 73 | ||
AK166030.1 | - | 1 - 2 | 230 | 73 | ||
AK171641.1 | - | 3 - 4 | 488 | 73 | ||
BC145805.1 | - | 3 - 4 | 578 | 40131108 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |