Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144527.3 | NP_653110.3 | 11 - 12 | 1954 | 72 | |
GenBank mRNA | AK030742.1 | - | 11 - 12 | 1919 | 72 | |
AK141103.1 | - | 11 - 12 | 1944 | 72 | ||
BC031729.1 | - | 11 - 12 | 1844 | 3992639 | 72 | |
BC114391.1 | - | 7 - 8 | 850 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |