Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144527.3 | NP_653110.3 | 5 - 6 | 1189 | 74 | |
GenBank mRNA | AK030742.1 | - | 5 - 6 | 1154 | 74 | |
AK085545.1 | - | 5 - 6 | 1136 | 74 | ||
AK141103.1 | - | 5 - 6 | 1179 | 74 | ||
BC008221.1 | - | 5 - 6 | 1141 | 74 | ||
BC031729.1 | - | 5 - 6 | 1079 | 3992639 | 74 | |
BC114391.1 | - | 1 - 2 | 85 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |