Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144527.3 | NP_653110.3 | 8 - 9 | 1651 | 76 | |
GenBank mRNA | AK030742.1 | - | 8 - 9 | 1616 | 76 | |
AK141103.1 | - | 8 - 9 | 1641 | 76 | ||
BC031729.1 | - | 8 - 9 | 1541 | 3992639 | 76 | |
BC114391.1 | - | 4 - 5 | 547 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |