Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010167.4 | NP_034297.2 | 15 - 16 | 1640 | 95 | |
GenBank mRNA | AJ007997.1 | - | 1 - 2 | 67 | 95 | |
AK041625.1 | - | 15 - 16 | 1641 | 95 | ||
BC120898.1 | - | 15 - 16 | 1624 | 40061597 | 95 | |
Y17115.1 | - | 16 - 17 | 1751 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |