Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172416.3 | NP_766004.1 | 4 - 5 | 859 | 67 | |
GenBank mRNA | AF533890.1 | - | 4 - 5 | 858 | 67 | |
AK004546.1 | - | 4 - 5 | 836 | 67 | ||
AK044897.1 | - | 4 - 5 | 858 | 67 | ||
AK050513.1 | - | 4 - 5 | 833 | 67 | ||
BC057635.1 | - | 4 - 5 | 797 | 4910872 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |