Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145743.2 | NP_665686.2 | 6 - 7 | 913 | 115 | |
GenBank mRNA | AF397909.1 | - | 5 - 6 | 616 | 115 | |
AK044595.1 | - | 6 - 7 | 789 | 115 | ||
AK170996.1 | - | 6 - 7 | 764 | 115 | ||
BC023308.1 | - | 5 - 6 | 590 | 115 | ||
BC089595.1 | - | 6 - 7 | 763 | 30118411 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |