Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081054.2 | NP_001074523.1 | 3 - 4 | 385 | 85 | |
GenBank mRNA | AK012351.1 | - | 3 - 4 | 386 | 85 | |
AK038801.1 | - | 3 - 4 | 394 | 85 | ||
AK041347.1 | - | 3 - 4 | 390 | 85 | ||
AK170225.1 | - | 3 - 4 | 330 | 85 | ||
BC070459.1 | - | 3 - 4 | 376 | 30605375 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |