Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081054.2 | NP_001074523.1 | 6 - 7 | 829 | 89 | |
GenBank mRNA | AK012351.1 | - | 6 - 7 | 830 | 89 | |
AK038801.1 | - | 6 - 7 | 838 | 89 | ||
AK041347.1 | - | 6 - 7 | 834 | 89 | ||
AK170225.1 | - | 6 - 7 | 774 | 89 | ||
BC037122.1 | - | 1 - 2 | 173 | 89 | ||
BC070459.1 | - | 6 - 7 | 820 | 30605375 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |