Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_001243074.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_001243074.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_001243075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_001243075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_026643.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_026643.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_029132.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
NM_029132.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014674.mm.1 |
TC1000002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014683.mm.1 |
NM_001243074.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014683.mm.1 |
NM_001243074.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014683.mm.1 |
TC1000002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
NM_001243074.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
NM_001243074.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
NM_001243075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
NM_001243075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
NM_029132.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
NM_029132.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014680.mm.1 |
TC1000002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
NM_001243074.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
NM_001243074.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
NM_001243075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
NM_001243075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
NM_029132.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
NM_029132.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014682.mm.1 |
TC1000002056.mm.1 |
MTA TC |