Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009185.3 | NP_033211.2 | 8 - 9 | 1143 | 104 | |
GenBank mRNA | AK049223.1 | - | 8 - 9 | 1080 | 104 | |
AK049550.1 | - | 8 - 9 | 1080 | 104 | ||
BC049865.1 | - | 8 - 9 | 1097 | 104 | ||
BC060706.1 | - | 8 - 9 | 1059 | 104 | ||
BC087545.1 | - | 7 - 8 | 834 | 104 | ||
BC108373.1 | - | 7 - 8 | 811 | 104 | ||
BC141175.1 | - | 8 - 9 | 1016 | 9055787 | 104 | |
BC145493.1 | - | 8 - 9 | 1016 | 9054056 | 104 | |
U36778.1 | - | 8 - 9 | 1061 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |