Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114333.2 | NP_001107805.1 | 2 - 3 | 1103 | 77 | |
NM_016976.3 | NP_058672.1 | 2 - 3 | 1103 | 77 | ||
GenBank mRNA | AF320126.2 | - | 2 - 3 | 1128 | 77 | |
BC067057.1 | - | 2 - 3 | 1077 | 30544252 | 77 | |
BC079566.1 | - | 1 - 2 | 1120 | 30536724 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |