Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011902 |
BC066795.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011902 |
AY355342.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011902 |
NM_172481.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011902 |
AK054426.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
695490 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
155073 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
751630 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
264537 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685802 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
820876 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946846 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
AF119253.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
AF119254.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
AK002532.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
BC019721.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
BC031711.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
K01922.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
K01926.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
M11357.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
M21931.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
NM_010378.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
V00832.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431279 |
X52643.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916654 |
AK017250.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916654 |
AK017424.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916654 |
BC087927.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916654 |
NM_009664.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916654 |
U65021.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431947 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
83723 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
961747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
336331 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685560 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493697 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
519698 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
199389 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
106798 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
86517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
168313 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P147988 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P103045 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011902 |
BC066795.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011902 |
AY355342.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011902 |
NM_172481.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011902 |
AK054426.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966950 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966954 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966931 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966939 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966960 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966948 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966946 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966949 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966938 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966947 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966961 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966965 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966943 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966935 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966952 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966929 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966932 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966956 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966963 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966969 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966927 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966941 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966936 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966962 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966951 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24966968 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01478590 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P147988 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P103045 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06122463 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06122507 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06122522 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P572679 |
BC066795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P572679 |
AY355342.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P572679 |
NM_172481.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P572679 |
AK054426.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P287790 |
BC066795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P287790 |
AY355342.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P287790 |
NM_172481.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P287790 |
AK054426.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134543 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135052 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P147988 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P103045 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06517384 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06446172 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06446163 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06584818 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06638581 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06446166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24966943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24967098 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966941 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966983 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966944 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966975 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966961 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06446170 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966951 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966963 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966929 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06463327 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966946 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966976 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966956 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966987 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06446172 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966942 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966935 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966977 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966984 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06584819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06639758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966948 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966938 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966928 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966927 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966965 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966967 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966960 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966943 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966980 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966969 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966926 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05933592 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24966970 |
- |
- |