Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013807.2 | NP_038835.2 | 6 - 7 | 844 | 71 | |
GenBank mRNA | AK156754.1 | - | 6 - 7 | 856 | 71 | |
BC031180.1 | - | 6 - 7 | 836 | 4988360 | 71 | |
BC063051.1 | - | 6 - 7 | 844 | 5718130 | 71 | |
U21392.1 | - | 6 - 7 | 691 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |