Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029988.mm.1 |
TC0400003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029988.mm.1 |
NM_001122992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029988.mm.1 |
NM_001122992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029988.mm.1 |
NM_020273.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029988.mm.1 |
NM_020273.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029996.mm.1 |
TC0400003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029996.mm.1 |
NM_001122992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029996.mm.1 |
NM_001122992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029996.mm.1 |
NM_020273.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029996.mm.1 |
NM_020273.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029991.mm.1 |
TC0400003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029991.mm.1 |
NM_001122992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029991.mm.1 |
NM_001122992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029989.mm.1 |
TC0400003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029989.mm.1 |
NM_001122992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029989.mm.1 |
NM_001122992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029989.mm.1 |
NM_020273.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029989.mm.1 |
NM_020273.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029987.mm.1 |
TC0400003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029987.mm.1 |
NM_001122992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029987.mm.1 |
NM_001122992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029987.mm.1 |
NM_020273.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029987.mm.1 |
NM_020273.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029994.mm.1 |
TC0400003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029994.mm.1 |
NM_001122992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029994.mm.1 |
NM_001122992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029994.mm.1 |
NM_020273.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029994.mm.1 |
NM_020273.2 |
Refseq |