Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025579.3 | NP_079855.2 | 1 - 2 | 107 | 96 | |
GenBank mRNA | AK007811.1 | - | 1 - 2 | 70 | 96 | |
AK013133.1 | - | 1 - 2 | 70 | 96 | ||
BC019668.1 | - | 1 - 2 | 52 | 3493047 | 96 | |
BY069348.1 | - | 1 - 2 | 108 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |