Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029937.2 | NP_001025108.1 | 1 - 2 | 149 | 56 | |
GenBank mRNA | BC138982.1 | - | 1 - 2 | 149 | 8862004 | 56 |
BC138985.1 | - | 1 - 2 | 149 | 8862007 | 56 | |
BC145485.1 | - | 1 - 2 | 149 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |