Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271730.1 | NP_001258659.1 | 2 - 3 | 658 | 79 | |
NM_013842.3 | NP_038870.2 | 2 - 3 | 658 | 79 | ||
GenBank mRNA | AF027963.3 | - | 2 - 3 | 332 | 79 | |
AF443192.1 | - | 2 - 3 | 658 | 79 | ||
AK007724.1 | - | 2 - 3 | 319 | 79 | ||
BC008153.1 | - | 2 - 3 | 273 | 79 | ||
BC016079.1 | - | 2 - 3 | 623 | 4489182 | 79 | |
BC029197.1 | - | 2 - 3 | 313 | 2648085 | 79 | |
BG800705.1 | - | 2 - 3 | 289 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |