Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009622.1 | NP_033752.1 | 8 - 9 | 1714 | 57 | |
GenBank mRNA | AK220469.1 | - | 2 - 3 | 185 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |