Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194052.3 | NP_918941.1 | 1 - 2 | 791 | 100 | |
GenBank mRNA | AK034902.1 | - | 1 - 2 | 735 | 100 | |
AY102281.1 | - | 1 - 2 | 748 | 100 | ||
AY114153.1 | - | 2 - 3 | 619 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |