Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029385.mm.1 |
TC0400003602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029385.mm.1 |
NM_001162433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029385.mm.1 |
NM_001162433.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
TC0400003602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
NM_001162432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
NM_001162432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
NM_001162433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
NM_001162433.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
NM_010693.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029379.mm.1 |
NM_010693.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029388.mm.1 |
TC0400003602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029388.mm.1 |
NM_001162433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029388.mm.1 |
NM_001162433.1 |
Refseq |