Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162432.1 | NP_001155904.1 | 6 - 7 | 568 | 84 | |
NM_001162433.1 | NP_001155905.1 | 6 - 7 | 610 | 84 | ||
NM_010693.3 | NP_034823.1 | 5 - 6 | 665 | 84 | ||
GenBank mRNA | AK088001.1 | - | 5 - 6 | 666 | 84 | |
BC011474.1 | - | 6 - 7 | 575 | 4191610 | 84 | |
CA577097.1 | - | 3 - 4 | 301 | 84 | ||
M12056.1 | - | 5 - 6 | 641 | 84 | ||
X03533.1 | - | 5 - 6 | 671 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |